missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorotoluene, 98%, Thermo Scientific™

Catalog Number p-106312
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19398-61-9
MFCD00000609
KFAKZJUYBOYVKA-UHFFFAOYSA-N
2,5-dichlorotoluene, benzene, 1,4-dichloro-2-methyl, toluene, 2,5-dichloro, 1,4-dichloro-2-methyl-benzene, toluene,5-dichloro, pubchem3700, acmc-209exk, ksc490m7d, benzene,4-dichloro-2-methyl
29572
1,4-dichloro-2-methylbenzene
CC1=C(C=CC(=C1)Cl)Cl
This item is not returnable. View return policy
19398-61-9
MFCD00000609
KFAKZJUYBOYVKA-UHFFFAOYSA-N
2,5-dichlorotoluene, benzene, 1,4-dichloro-2-methyl, toluene, 2,5-dichloro, 1,4-dichloro-2-methyl-benzene, toluene,5-dichloro, pubchem3700, acmc-209exk, ksc490m7d, benzene,4-dichloro-2-methyl
29572
1,4-dichloro-2-methylbenzene
CC1=C(C=CC(=C1)Cl)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2500g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H6Cl2
Linear Formula CH3C6H3Cl2
Refractive Index 1.5460 to 1.5480 (20°C, 589nm)
Quantity 5 g
Beilstein 05, 296
Molecular Weight (g/mol) 161.025
Formula Weight 161.03
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2, 5-Dichlorotoluene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'