missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloropyridine-3-carbonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 210.45 |
Percent Purity | 97% |
Chemical Name or Material | 2, 5-Dichloropyridine-3-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'