missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-dichlorophenyl isothiocyanate, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
Assay Percent Range | 97+% |
Quantity | 10 g |
Formula Weight | 204.08 |
Percent Purity | 97% |
Chemical Name or Material | 2, 5-Dichlorophenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'