missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7045782
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10541-69-2
C7H7Cl2N
176.04
MFCD00052391
AKGJLIXNRPNPCH-UHFFFAOYSA-N
2,5-dichlorobenzylamine, 2,5-dichlorophenyl methanamine, benzenemethanamine, 2,5-dichloro, 2,5-dichloro-benzylamine, 1-2,5-dichlorophenyl methanamine, 2,5-dichlorophenyl methylamine, 2,5-dichlorobenzylam, acmc-1bxya, rarechem al bw 0384, 2,5-dichlorobenzyl amine
457600
(2,5-dichlorophenyl)methanamine
C1=CC(=C(C=C1Cl)CN)Cl
This item is not returnable. View return policy
10541-69-2
C7H7Cl2N
176.04
MFCD00052391
AKGJLIXNRPNPCH-UHFFFAOYSA-N
2,5-dichlorobenzylamine, 2,5-dichlorophenyl methanamine, benzenemethanamine, 2,5-dichloro, 2,5-dichloro-benzylamine, 1-2,5-dichlorophenyl methanamine, 2,5-dichlorophenyl methylamine, 2,5-dichlorobenzylam, acmc-1bxya, rarechem al bw 0384, 2,5-dichlorobenzyl amine
457600
(2,5-dichlorophenyl)methanamine
C1=CC(=C(C=C1Cl)CN)Cl

2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Incompatible with strong oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY
Boiling Point 74°C to 76°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Quantity 1 g
UN Number UN2735
Beilstein 2639302
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'