missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzhydrazide, 98+%, Thermo Scientific™

Catalog Number p-7022147
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
67487-35-8
C7H6Cl2N2O
205.04
MFCD00014754
XECBCXNWGBDIMG-UHFFFAOYSA-N
2,5-dichlorobenzhydrazide, 1,4-dichlorobenzene-2-carbohydrazide, #, 2,5-dichlorobenzoic hydrazide, 2,5-bis chloranyl benzohydrazide, labotest-bb lt00454263, labotest-bb lt03497761, 2,5-dichlorobenzoic acid hydrazide
144246
2,5-dichlorobenzohydrazide
NNC(=O)C1=CC(Cl)=CC=C1Cl
This item is not returnable. View return policy
67487-35-8
C7H6Cl2N2O
205.04
MFCD00014754
XECBCXNWGBDIMG-UHFFFAOYSA-N
2,5-dichlorobenzhydrazide, 1,4-dichlorobenzene-2-carbohydrazide, #, 2,5-dichlorobenzoic hydrazide, 2,5-bis chloranyl benzohydrazide, labotest-bb lt00454263, labotest-bb lt03497761, 2,5-dichlorobenzoic acid hydrazide
144246
2,5-dichlorobenzohydrazide
NNC(=O)C1=CC(Cl)=CC=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 181°C
Quantity 5 g
Beilstein 4394418
Formula Weight 205.04
Percent Purity ≥98%
Chemical Name or Material 2,5-Dichlorobenzhydrazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'