missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-104275
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6361-23-5
C7H4Cl2O
175.01
MFCD00156140
BUXHYMZMVMNDMG-UHFFFAOYSA-N
2,5-dichloro-benzaldehyde, 2,5-dichlorobenzaldehdye, benzaldehyde, 2,5-dichloro, pubchem3081, 2.5-dichlorbenzaldehyd, 2,5-dichlorbenzaldehyde, acmc-1bkcw, 2,5-dichloro benzaldehdye, ksc493o7f, 2,5-dichlorobenzaldehyde
80702
2,5-dichlorobenzaldehyde
ClC1=CC=C(Cl)C(C=O)=C1
This item is not returnable. View return policy
6361-23-5
C7H4Cl2O
175.01
MFCD00156140
BUXHYMZMVMNDMG-UHFFFAOYSA-N
2,5-dichloro-benzaldehyde, 2,5-dichlorobenzaldehdye, benzaldehyde, 2,5-dichloro, pubchem3081, 2.5-dichlorbenzaldehyd, 2,5-dichlorbenzaldehyde, acmc-1bkcw, 2,5-dichloro benzaldehdye, ksc493o7f, 2,5-dichlorobenzaldehyde
80702
2,5-dichlorobenzaldehyde
ClC1=CC=C(Cl)C(C=O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 58°C
Color White
Boiling Point 231°C to 233°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(Cl)CHO
Quantity 1 g
Formula Weight 175.02
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dichlorobenzaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichlorobenzaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'