missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloro-4,6-dimethylnicotinonitrile, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2,5-dichloro-4,6-dimethylnicotinonitrile, 3-pyridinecarbonitrile, 2,5-dichloro-4,6-dimethyl, 2,5-dichloro-4,6-dimethyl-nicotinonitrile, pubchem20434, ksc669q6f, 2,5-dichloro-3-cyano-4,6-dimethylpyridine, 2,5-dichloro-4,6-dimethyl-3-pyridyl cyanide, 3-pyridinecarbonitrile,5-dichloro-4,6-dimethyl, 2,5-dichloro-4,6-dimethyl-pyridine-3-carbonitrile, 3-pyridinecarbonitrile,2,5-dichloro-4,6-dimethyl |
Formula Weight | 201.06 |
Percent Purity | 95% |
Chemical Name or Material | 2, 5-Dichloro-4, 6-dimethylnicotinonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'