missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-3-nitropyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-3211526
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
21427-62-3
C5H2Cl2N2O2
192.99
MFCD06658963
OBUGJYJQJWMOQO-UHFFFAOYSA-N
2,5-dichloro-3-nitro-pyridine, pyridine, 2,5-dichloro-3-nitro, 2.5-dichloro-3-nitropyridine, 2,5-dichloro-3-nitro pyridine, pubchem2267, acmc-1cdlk, 2,5dichloro-3-nitropyridine, ksc495e7t, 2,5 dichloro-3-nitropyridine, 2,5 dichloro-3-nitro pyridine
8063331
2,5-dichloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
21427-62-3
C5H2Cl2N2O2
192.99
MFCD06658963
OBUGJYJQJWMOQO-UHFFFAOYSA-N
2,5-dichloro-3-nitro-pyridine, pyridine, 2,5-dichloro-3-nitro, 2.5-dichloro-3-nitropyridine, 2,5-dichloro-3-nitro pyridine, pubchem2267, acmc-1cdlk, 2,5dichloro-3-nitropyridine, ksc495e7t, 2,5 dichloro-3-nitropyridine, 2,5 dichloro-3-nitro pyridine
8063331
2,5-dichloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 39°C to 45°C
Color Beige-Green to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20, IV, 2532
Formula Weight 192.99
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2, 5-Dichloro-3-nitropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichloro-3-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'