Alfa Aesar™ 2,5-Dichloro-3-nitropyridine, ≥97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64276.14

25GR 2,5-Dichloro-3-nitropyridine, 97+% 25g

UNSPSC: 12352005

Code: NEW

Additional Details:
Additional Details: CAS Number: 21427-62-3 Weight: 0.02500kg


Quantity 25g

Product Code. 15411399

Quantity Price
1 £ 77.59 / 25g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2,5-Dichloro-3-nitropyridine
CAS 21427-62-3
Formula Weight 192.98
MDL Number MFCD06658963
UN Number UN2811
Beilstein 147539

Specification

Formula Weight 192.98
Melting Point 41° to 45°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,5-Dichloro-3-nitropyridine
Physical Form Solid
Assay ≥97%
MDL Number MFCD06658963
CAS 21427-62-3
UN Number UN2811
Beilstein 147539
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 25g
Safety and Handling

Danger

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.