Learn More
Thermo Scientific Maybridge 2,5-Dibromoterephthalic acid, 1g, 97%
Brand: Thermo Scientific Maybridge BTB11564DA
2,5-Dibromoterephthalic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 13731-82-3 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge BTB11564DA
2,5-Dibromoterephthalic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 13731-82-3 Weight : 0.05100kg