missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog Number 15468548
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
15862-37-0
C5H2Br2N2O2
281.891
MFCD09266223
OQKWPJCAKRVADO-UHFFFAOYSA-N
2,5-dibromo-3-nitro-pyridine, 2,5-di bromo-3-nitro pyridine, pyridine, 2,5-dibromo-3-nitro, pubchem5505, acmc-1bp61, ksc495e2t, 2 pound not5-dibromo-3-nitropyridine
298470
2,5-dibromo-3-nitropyridine
C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]
This item is not returnable. View return policy
15862-37-0
C5H2Br2N2O2
281.891
MFCD09266223
OQKWPJCAKRVADO-UHFFFAOYSA-N
2,5-dibromo-3-nitro-pyridine, 2,5-di bromo-3-nitro pyridine, pyridine, 2,5-dibromo-3-nitro, pubchem5505, acmc-1bp61, ksc495e2t, 2 pound not5-dibromo-3-nitropyridine
298470
2,5-dibromo-3-nitropyridine
C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 5 g
Formula Weight 281.89
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,5-Dibromo-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'