missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,5-Dibromo-3-n-dodecylthiophene, 97%

Brand:  Alfa Aesar™ H55958.06

Code : NEW

Additional Details : CAS Number : 148256-63-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15402368

  • £270.00 / 5g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
2,5-Dibromo-3-n-dodecylthiophene is used as an OLED intermediate, flavor and fragrance intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

148256-63-7
410.252
ZIZGNWAMVWNAJT-UHFFFAOYSA-N
5072754
CCCCCCCCCCCCC1=C(SC(=C1)Br)Br
C16H26Br2S
MFCD00274315
thiophene, 2,5-dibromo-3-dodecyl, 2,5-dibromo-3-laurylthiophene, 2,5-dibromo-3-dodecyl-thiophene, 2,5-bromo-3-dodecylthiophene, acmc-209cip, ksc496a6t, bidd:gt0375, thiophene,2,5-dibromo-3-dodecyl, 2,5-bis bromanyl-3-dodecyl-thiophene, 2,5-dibromo-3-dodecylthiophene
2,5-dibromo-3-dodecylthiophene

Specifications

2,5-Dibromo-3-n-dodecylthiophene
>113°C (235°F)
135°C (2.5mmHg)
148256-63-7
MFCD00274315
Slightly soluble in water.
CCCCCCCCCCCCC1=C(SC(=C1)Br)Br
410.252
410.25
1.321
1.53
5g
C16H26Br2S
thiophene, 2,5-dibromo-3-dodecyl, 2,5-dibromo-3-laurylthiophene, 2,5-dibromo-3-dodecyl-thiophene, 2,5-bromo-3-dodecylthiophene, acmc-209cip, ksc496a6t, bidd:gt0375, thiophene,2,5-dibromo-3-dodecyl, 2,5-bis bromanyl-3-dodecyl-thiophene, 2,5-dibromo-3-dodecylthiophene
ZIZGNWAMVWNAJT-UHFFFAOYSA-N
2,5-dibromo-3-dodecylthiophene
5072754
97%
Videos
SDS Documents
Certificates