missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3,4-dinitrothiophene, Thermo Scientific Chemicals

Catalog Number 13488498
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
52431-30-8
C4Br2N2O4S
331.92
MFCD00015537
AHGHPBPARMANQD-UHFFFAOYSA-N
2,5-dibromo-3,4-dinitro-thiophene, thiophene, 2,5-dibromo-3,4-dinitro, acmc-1apgk, ksc491i5j, 2,5-dibromo-3,4-dinitrothiophene gc
257024
2,5-dibromo-3,4-dinitrothiophene
[O-][N+](=O)C1=C(Br)SC(Br)=C1[N+]([O-])=O
This item is not returnable. View return policy
52431-30-8
C4Br2N2O4S
331.92
MFCD00015537
AHGHPBPARMANQD-UHFFFAOYSA-N
2,5-dibromo-3,4-dinitro-thiophene, thiophene, 2,5-dibromo-3,4-dinitro, acmc-1apgk, ksc491i5j, 2,5-dibromo-3,4-dinitrothiophene gc
257024
2,5-dibromo-3,4-dinitrothiophene
[O-][N+](=O)C1=C(Br)SC(Br)=C1[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 132.0°C to 134.0°C
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 331.94
Chemical Name or Material 2, 5-Dibromo-3, 4-dinitrothiophene
compliance-icons
Product Identifier
  • 2,5-Dibromo-3,4-dinitrothiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'