missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di-tert-butyl-p-benzoquinone, 99%, Thermo Scientific Chemicals

Catalog Number p-7058338
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2460-77-7
C14H20O2
220.312
MFCD00019442
ZZYASVWWDLJXIM-UHFFFAOYSA-N
2,5-di-tert-butyl-1,4-benzoquinone, 2,5-di-tert-butylquinone, 2,5-di-tert-butylsemiquinone, 2,5-di-tert-butylbenzoquinone, 2,5-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,5-di-tert-butyl-p-quinone, 2,5-di-tert-butyl-p-benzoquinone, unii-g893osv02d, p-benzoquinone, 2,5-di-tert-butyl, 2,5-di-t-butyl-p-benzoquinone
17161
2,5-ditert-butylcyclohexa-2,5-diene-1,4-dione
CC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)C
This item is not returnable. View return policy
2460-77-7
C14H20O2
220.312
MFCD00019442
ZZYASVWWDLJXIM-UHFFFAOYSA-N
2,5-di-tert-butyl-1,4-benzoquinone, 2,5-di-tert-butylquinone, 2,5-di-tert-butylsemiquinone, 2,5-di-tert-butylbenzoquinone, 2,5-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,5-di-tert-butyl-p-quinone, 2,5-di-tert-butyl-p-benzoquinone, unii-g893osv02d, p-benzoquinone, 2,5-di-tert-butyl, 2,5-di-t-butyl-p-benzoquinone
17161
2,5-ditert-butylcyclohexa-2,5-diene-1,4-dione
CC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 152°C to 153°C
Quantity 5 g
Beilstein 2047945
Formula Weight 220.31
Percent Purity 99%
Chemical Name or Material 2,5-Di-tert-butyl-p-benzoquinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'