missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
IUPAC Name | 2,5-bis(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride |
Formula Weight | 372.67 |
Percent Purity | 97% |
Chemical Name or Material | 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'