missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di(2,2,2-trifluoroethoxy)benzene-1-carbohydrazide, ≥95%, Thermo Scientific™

Catalog Number p-2697126
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
50778-75-1
C11H10F6N2O3
332.202
MFCD00221192
WJYCMPVGQTYGSQ-UHFFFAOYSA-N
2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzohydrazide, 2,5-di 2,2,2-trifluoroethoxy benzene-1-carbohydrazide, maybridge1_008849, 2,5-bis 2,2,2-tris fluoranyl ethoxy benzohydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzenecarbohydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazi, 2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazide 95+%, 2,5-bis 2,2,2-trifluoroethoxy benzoic acidhydrazide
3547540
2,5-bis(2,2,2-trifluoroethoxy)benzohydrazide
C1=CC(=C(C=C1OCC(F)(F)F)C(=O)NN)OCC(F)(F)F
This item is not returnable. View return policy
50778-75-1
C11H10F6N2O3
332.202
MFCD00221192
WJYCMPVGQTYGSQ-UHFFFAOYSA-N
2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzohydrazide, 2,5-di 2,2,2-trifluoroethoxy benzene-1-carbohydrazide, maybridge1_008849, 2,5-bis 2,2,2-tris fluoranyl ethoxy benzohydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzenecarbohydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazi, 2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazide 95+%, 2,5-bis 2,2,2-trifluoroethoxy benzoic acidhydrazide
3547540
2,5-bis(2,2,2-trifluoroethoxy)benzohydrazide
C1=CC(=C(C=C1OCC(F)(F)F)C(=O)NN)OCC(F)(F)F
CAS Min % 90.0
Assay Percent Range 90%+
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 332.2
Percent Purity 90%
Chemical Name or Material 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-carbohydrazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'