missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-tert-Butylphenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide, 95%

Brand:  Alfa Aesar™ H65545.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459726

  • £70.28 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-tert-Butylphenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
321.46
MFCD27975780

Specifications

2-(4-tert-Butylphenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
95%
MFCD27975780
Solid
321.46
1g
Videos
SDS