missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Nitrophenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62398.03

Code : NEW

Additional Details : CAS Number : 72678-85-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407386

  • £46.00 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Nitrophenyl)thiazolidine-4-carboxylic acid
254.26
257273
72678-85-4
MFCD00666983

Specifications

2-(4-Nitrophenyl)thiazolidine-4-carboxylic acid
Solid
254.26
1g
72678-85-4
97%
MFCD00666983
257273
Videos
SDS