missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Nitrophenyl)ethylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.
Solubility
Soluble in methanol almost transparency.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.
Solubility
Soluble in methanol almost transparency.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 200°C (literature) |
Quantity | 25 g |
Beilstein | 3568915 |
Solubility Information | Soluble in methanol almost transparency. |
Formula Weight | 202.64 |
Percent Purity | ≥98% |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2-(4-Nitrophenyl)ethylamine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'