missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Nitrophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64374.03

Code : NEW

Additional Details : CAS Number : 729-13-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451549

  • £119.78 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Nitrophenyl)benzimidazole
729-13-5
239.23
MFCD00225481

Specifications

2-(4-Nitrophenyl)benzimidazole
Solid
239.23
1g
729-13-5
95%
MFCD00225481
Videos
SDS