Alfa Aesar™ 2-(4-Nitrophenyl)benzimidazole, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64374.06

UNSPSC: 12352005

Code: NEW

Additional Details:
Additional Details: CAS Number: 729-13-5 Weight: 0.00500kg


Quantity 5g

Product Code. 15461549

Quantity Price
1 £ 597.84 / 5g
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-(4-Nitrophenyl)benzimidazole
CAS 729-13-5
Formula Weight 239.23
MDL Number MFCD00225481

Specification

Formula Weight 239.23
Quantity 5g
Chemical Name or Material 2-(4-Nitrophenyl)benzimidazole
Physical Form Solid
CAS 729-13-5
MDL Number MFCD00225481
Assay 95%