missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-[4-(Methylthio)phenyl]benzimidazole, 95%

Brand:  Alfa Aesar™ H64661.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15481949

  • £62.96 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

2-[4-(Methylthio)phenyl]benzimidazole
240.32
MFCD06090773

Specifications

2-[4-(Methylthio)phenyl]benzimidazole
Solid
240.32
1g
95%
MFCD06090773
Videos
SDS