missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Methylphenyl)benzimidazole, 97%

Brand:  Alfa Aesar™ H58894.04

Code : NEW

Additional Details : CAS Number : 120-03-6 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 15455078

  • £144.10 / 2g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Methylphenyl)benzimidazole
120-03-6
208.26
MFCD00209485

Specifications

2-(4-Methylphenyl)benzimidazole
120-03-6
97%
MFCD00209485
Pale brown
2g
Solid
208.26
265° to 269°C
Videos
SDS