missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)benzimidazole, 97%, Thermo Scientific™

Catalog Number p-7053060
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
120-03-6
C14H12N2
208.264
MFCD00209485
YJSWQFYMPODDTQ-UHFFFAOYSA-N
2-p-tolyl-1h-benzoimidazole, 2-4-methylphenyl-1h-benzimidazole, 2-4-methylphenyl benzimidazole, 2-p-tolyl-1h-benzo d imidazole, 1h-benzimidazole,2-4-methylphenyl, 2-4-methylphenyl-1h-1,3-benzodiazole, 1h-benzimidazole, 2-4-methylphenyl, 2-ptolyl-1h-benzoimidazole, 2-4-methylphenyl-1h-benzo d imidazole
725982
2-(4-methylphenyl)-1H-benzimidazole
CC1=CC=C(C=C1)C2=NC3=CC=CC=C3N2
This item is not returnable. View return policy
120-03-6
C14H12N2
208.264
MFCD00209485
YJSWQFYMPODDTQ-UHFFFAOYSA-N
2-p-tolyl-1h-benzoimidazole, 2-4-methylphenyl-1h-benzimidazole, 2-4-methylphenyl benzimidazole, 2-p-tolyl-1h-benzo d imidazole, 1h-benzimidazole,2-4-methylphenyl, 2-4-methylphenyl-1h-1,3-benzodiazole, 1h-benzimidazole, 2-4-methylphenyl, 2-ptolyl-1h-benzoimidazole, 2-4-methylphenyl-1h-benzo d imidazole
725982
2-(4-methylphenyl)-1H-benzimidazole
CC1=CC=C(C=C1)C2=NC3=CC=CC=C3N2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 265°C to 269°C
Color Brown
Quantity 2 g
Formula Weight 208.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(4-Methylphenyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'