missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methylperhydro-1,4-diazepin-1-yl)benzylamine, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 219.33 |
Percent Purity | 90% |
Chemical Name or Material | 2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'