missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)piperazine, 95%, Thermo Scientific™

Catalog Number 15403477
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 91517-26-9
Melting Point 55°C to 63°C
Molecular Formula C11H16N2O
MDL Number MFCD05864677
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Synonym 2-4-methoxyphenyl piperazine, piperazine, 2-4-methoxyphenyl, piperazine,2-4-methoxyphenyl, 2-4-methoxypheny-piperazine, 2-4-methoxyphenyl-piperazine, 2-4-methoxy-phenyl-piperazine, 4-methoxy-1-piperazin-2-ylbenzene, rs-2-4-methoxyphenyl piperazine, rs-3-4-methoxyphenyl piperazine, 1-methoxy-4-piperazin-2-ylbenzene
InChI Key INKLSJITWMAFRT-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C2CNCCN2
IUPAC Name 2-(4-methoxyphenyl)piperazine
Molecular Weight (g/mol) 192.262
PubChem CID 2771753
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 2-(4-Methoxyphenyl)piperazine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'