missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methoxyphenoxy)ethylamine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Linear Formula | NH2(CH2)2OC6H4OCH3 |
Quantity | 1 g |
Formula Weight | 167.21 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 2-(4-Methoxyphenoxy)ethylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'