missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Especificaciones
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 213.19 |
Percent Purity | 97% |
Chemical Name or Material | 2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2, 3-dihydro-1H-pyrrol-3-yl]acetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'