missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid, 97%, Thermo Scientific™

Catalog Number p-2697582
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
77978-73-5
C9H11NO5
213.189
MFCD00173859
WLKUVVIAHUXPSB-UHFFFAOYSA-N
2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3-yl acetic acid, 4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 1h-pyrrole-3-aceticacid, 2,3-dihydro-4-methoxycarbonyl-5-methyl-2-oxo, maybridge1_002247, 3-methoxycarbonyl-2-methyl-5-oxo-2-pyrrolin-4-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3yl acetic acid
2774998
2-(4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl)acetic acid
CC1=C(C(C(=O)N1)CC(=O)O)C(=O)OC
This item is not returnable. View return policy
77978-73-5
C9H11NO5
213.189
MFCD00173859
WLKUVVIAHUXPSB-UHFFFAOYSA-N
2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3-yl acetic acid, 4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 1h-pyrrole-3-aceticacid, 2,3-dihydro-4-methoxycarbonyl-5-methyl-2-oxo, maybridge1_002247, 3-methoxycarbonyl-2-methyl-5-oxo-2-pyrrolin-4-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3yl acetic acid
2774998
2-(4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl)acetic acid
CC1=C(C(C(=O)N1)CC(=O)O)C(=O)OC
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 213.19
Percent Purity 97%
Chemical Name or Material 2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2, 3-dihydro-1H-pyrrol-3-yl]acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'