missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Isopropylphenyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62644.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429148

  • £59.78 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Isopropylphenyl)thiazolidine
207.34
MFCD16685185

Specifications

2-(4-Isopropylphenyl)thiazolidine
Solid
207.34
1g
97%
MFCD16685185
Videos
SDS