missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Isopropylphenyl)-5-nitrobenzimidazole, 95%

Brand:  Alfa Aesar™ H64818.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15492189

  • £85.44 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Isopropylphenyl)-5-nitrobenzimidazole
281.32
MFCD17779818

Specifications

2-(4-Isopropylphenyl)-5-nitrobenzimidazole
Solid
281.32
1g
95%
MFCD17779818
Videos
SDS