missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isocyanatophenyl)-5-(trifluoromethyl)pyridine, ≥97%, Thermo Scientific™

Catalog Number p-3586625
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
906352-73-6
C13H7F3N2O
264.207
MFCD09817476
GFEVPIXAPFROGH-UHFFFAOYSA-N
2-4-isocyanatophenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl-2-pyridyl benzenisocyanate
24229533
2-(4-isocyanatophenyl)-5-(trifluoromethyl)pyridine
C1=CC(=CC=C1C2=NC=C(C=C2)C(F)(F)F)N=C=O
This item is not returnable. View return policy
906352-73-6
C13H7F3N2O
264.207
MFCD09817476
GFEVPIXAPFROGH-UHFFFAOYSA-N
2-4-isocyanatophenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl-2-pyridyl benzenisocyanate
24229533
2-(4-isocyanatophenyl)-5-(trifluoromethyl)pyridine
C1=CC(=CC=C1C2=NC=C(C=C2)C(F)(F)F)N=C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 264.21
Percent Purity 97%
Chemical Name or Material 2-(4-Isocyanatophenyl)-5-(trifluoromethyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'