missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride, 95%

Brand:  Alfa Aesar™ H65601.03

Code : NEW

Additional Details : CAS Number : 57591-61-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412579

  • £15.40 / 1g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride
217.65
(R)-Amino-(4-hydroxyphenyl)acetic acid methyl ester hydrochloride
57591-61-4
MFCD01862152

Specifications

2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride
Solid
217.65
(R)-Amino-(4-hydroxyphenyl)acetic acid methyl ester hydrochloride
1g
57591-61-4
95%
MFCD01862152
187° to 190°C
Videos
SDS