missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)acetamide, 97%, Maybridge™

Catalog Number p-2696251
Click to view available options
Quantity:
50 g
100 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
17194-82-0
C8H9NO2
151.165
YBPAYPRLUDCSEY-UHFFFAOYSA-N
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
86986
2-(4-hydroxyphenyl)acetamide
C1=CC(=CC=C1CC(=O)N)O
This item is not returnable. View return policy
17194-82-0
C8H9NO2
151.165
YBPAYPRLUDCSEY-UHFFFAOYSA-N
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
86986
2-(4-hydroxyphenyl)acetamide
C1=CC(=CC=C1CC(=O)N)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 2-(4-Hydroxyphenyl)acetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'