missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiyn-1,6-diol, 97%, Thermo Scientific Chemicals

Catalog Number 11307506
26.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3031-68-3
C6H6O2
110.112
MFCD00020619
JXMQYKBAZRDVTC-UHFFFAOYSA-N
2,4-hexadiyne-1,6-diol, diacetylene glycol, 2,4-hexadiynediol, 2,4-hexadiyn-1,6-diol, 2,6-diol, acmc-1ahii, hoch2c=c-c=cch2oh, 1,6-dihydroxy-2,4-hexadiyne, hexa-2, 4-diyne-1, 6-diol, jxmqykbazrdvtc-uhfffaoysa
94973
hexa-2,4-diyne-1,6-diol
C(C#CC#CCO)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3031-68-3
C6H6O2
110.112
MFCD00020619
JXMQYKBAZRDVTC-UHFFFAOYSA-N
2,4-hexadiyne-1,6-diol, diacetylene glycol, 2,4-hexadiynediol, 2,4-hexadiyn-1,6-diol, 2,6-diol, acmc-1ahii, hoch2c=c-c=cch2oh, 1,6-dihydroxy-2,4-hexadiyne, hexa-2, 4-diyne-1, 6-diol, jxmqykbazrdvtc-uhfffaoysa
94973
hexa-2,4-diyne-1,6-diol
C(C#CC#CCO)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 110°C to 114°C
Linear Formula HOCH2C≡C-C≡CCH2OH
Quantity 1 g
UN Number UN1325
Beilstein 773791
Formula Weight 110.11
Percent Purity 97%
Chemical Name or Material 2,4-Hexadiyn-1,6-diol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'