missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiene, 90%, Tech., mixture of isomers, Thermo Scientific Chemicals

Catalog Number p-141469
24.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
592-46-1
C6H10
82.15
MFCD00009297,MFCD00009297
APPOKADJQUIAHP-CIIODKQPSA-N
unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
643786
(2Z,4E)-hexa-2,4-diene
C\C=C\C=C/C
This item is not returnable. View return policy
592-46-1
C6H10
82.15
MFCD00009297,MFCD00009297
APPOKADJQUIAHP-CIIODKQPSA-N
unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
643786
(2Z,4E)-hexa-2,4-diene
C\C=C\C=C/C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 0.7200g/mL
Boiling Point 82°C
Flash Point -8°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCH=CHCH3
Refractive Index 1.45
Quantity 1 g
Beilstein 01, 254
Specific Gravity 0.72
Solubility Information Solubility in water: insoluble
Formula Weight 82.14
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2, 4-Hexadiene, tech. 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Hexadiene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'