missing translation for 'onlineSavingsMsg'
Learn More

2-[(4-Fluorophenyl)thio]acetic acid, 97%, Maybridge™

Brand:  Maybridge KM09206EE

Code : 31

Additional Details : CAS Number : 332-51-4 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 10001484

  • £125.00 / 50g

Estimated Shipment: 18-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

332-51-4
186.2
HQEROVXUKINLPI-UHFFFAOYSA-N
736197
C1=CC(=CC=C1F)SCC(=O)O
C8H7FO2S
MFCD00219825
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
2-(4-fluorophenyl)sulfanylacetic acid

Specifications

2-[(4-Fluorophenyl)thio]acetic acid
332-51-4
97%
FC6H4SCH2CO2H
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
C1=CC(=CC=C1F)SCC(=O)O
186.2
186.21
50g
97.0
C8H7FO2S
MFCD00219825
HQEROVXUKINLPI-UHFFFAOYSA-N
2-(4-fluorophenyl)sulfanylacetic acid
736197
97%
Videos
SDS Documents
Certificates