missing translation for 'onlineSavingsMsg'
Learn More

2-[(4-Fluorophenyl)sulfonyl]-N'-hydroxyethanimidamide, 97%, Thermo Scientific™

Catalog Number p-2706204
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175203-76-6
C8H9FN2O3S
232.229
MFCD00205110
STPCRDRACVTHTE-UHFFFAOYSA-N
2-4-fluorophenyl sulfonyl-n'-hydroxyethanimidamide, 2-4-fluorobenzenesulfonyl acetamide oxime, 2-4-fluorobenzenesulphonyl acetamide oxime, 2-4-fluorophenyl sulfonyl-n-hydroxyacetimidamide, 2-4-fluorophenyl sulfonyl-n-hydroxyethanimidamide, 1-4-fluorophenyl sulfonyl-2-hydroxyimino eth-2-ylamine, 2-4-fluorobenzenesulphonyl acetamidoxime, 2-4-fluorobenzenesulfonyl-n'-hydroxyethanimidamide, z-2-4-fluorobenzenesulfonyl-n'-hydroxyethanimidamide, 2-4-fluorophenylsulfonyl-n-hydroxy-acetamidine
9580384
2-(4-fluorophenyl)sulfonyl-N'-hydroxyethanimidamide
C1=CC(=CC=C1F)S(=O)(=O)CC(=NO)N
This item is not returnable. View return policy
175203-76-6
C8H9FN2O3S
232.229
MFCD00205110
STPCRDRACVTHTE-UHFFFAOYSA-N
2-4-fluorophenyl sulfonyl-n'-hydroxyethanimidamide, 2-4-fluorobenzenesulfonyl acetamide oxime, 2-4-fluorobenzenesulphonyl acetamide oxime, 2-4-fluorophenyl sulfonyl-n-hydroxyacetimidamide, 2-4-fluorophenyl sulfonyl-n-hydroxyethanimidamide, 1-4-fluorophenyl sulfonyl-2-hydroxyimino eth-2-ylamine, 2-4-fluorobenzenesulphonyl acetamidoxime, 2-4-fluorobenzenesulfonyl-n'-hydroxyethanimidamide, z-2-4-fluorobenzenesulfonyl-n'-hydroxyethanimidamide, 2-4-fluorophenylsulfonyl-n-hydroxy-acetamidine
9580384
2-(4-fluorophenyl)sulfonyl-N'-hydroxyethanimidamide
C1=CC(=CC=C1F)S(=O)(=O)CC(=NO)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula FC6H4SO2CH2C(=NOH)NH2
Quantity 1 g
Formula Weight 232.23
Percent Purity 97%
Chemical Name or Material 2-[(4-Fluorophenyl)sulfonyl]-N';-hydroxyethanimidamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'