missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Fluorophenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide, 95%

Brand:  Alfa Aesar™ H65990.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450077

  • £74.73 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Fluorophenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
283.35
MFCD08480187

Specifications

2-(4-Fluorophenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
Solid
283.35
1g
95%
MFCD08480187
Videos
SDS