missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Ethoxyphenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62533.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407546

  • £99.64 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Ethoxyphenyl)thiazolidine-4-carboxylic acid
253.32
MFCD01071049

Specifications

2-(4-Ethoxyphenyl)thiazolidine-4-carboxylic acid
Solid
253.32
1g
97%
MFCD01071049
Videos
SDS