missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyl-1,3-thiazole-5-sulfonyl chloride, ≥97%, Thermo Scientific™

Catalog Number p-3175139
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
868755-57-1
C15H10ClNO2S2
335.82
MFCD08271911
AWNJOBYLGNWNCQ-UHFFFAOYSA-N
diphenyl-1,3-thiazole-5-sulfonyl chloride, 2,4-diphenylthiazole-5-sulfonyl chloride, 5-thiazolesulfonylchloride, 2,4-diphenyl, 5-chlorosulphonyl-2,4-diphenyl-1,3-thiazole
18525757
ClS(=O)(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
868755-57-1
C15H10ClNO2S2
335.82
MFCD08271911
AWNJOBYLGNWNCQ-UHFFFAOYSA-N
diphenyl-1,3-thiazole-5-sulfonyl chloride, 2,4-diphenylthiazole-5-sulfonyl chloride, 5-thiazolesulfonylchloride, 2,4-diphenyl, 5-chlorosulphonyl-2,4-diphenyl-1,3-thiazole
18525757
ClS(=O)(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name diphenyl-1,3-thiazole-5-sulfonyl chloride
Formula Weight 335.83
Percent Purity 97%
Chemical Name or Material 2, 4-Diphenyl-1, 3-thiazole-5-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'