missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylstyrene, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number 15234694
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2234-20-0
C10H12
132.21
MFCD00014937
OEVVKKAVYQFQNV-UHFFFAOYSA-N
2,4-dimethylstyrene, 4-vinyl-m-xylene, styrene, 2,4-dimethyl, 1,3-dimethyl-4-vinylbenzene, 1-vinyl-2,4-dimethylbenzene, 1,3-dimethyl-4-ethenylbenzene, benzene, 1-ethenyl-2,4-dimethyl, unii-543es1o25e, styrene,4-dimethyl, acmc-1cb1s
16694
1-ethenyl-2,4-dimethylbenzene
CC1=CC(=C(C=C1)C=C)C
This item is not returnable. View return policy
2234-20-0
C10H12
132.21
MFCD00014937
OEVVKKAVYQFQNV-UHFFFAOYSA-N
2,4-dimethylstyrene, 4-vinyl-m-xylene, styrene, 2,4-dimethyl, 1,3-dimethyl-4-vinylbenzene, 1-vinyl-2,4-dimethylbenzene, 1,3-dimethyl-4-ethenylbenzene, benzene, 1-ethenyl-2,4-dimethyl, unii-543es1o25e, styrene,4-dimethyl, acmc-1cb1s
16694
1-ethenyl-2,4-dimethylbenzene
CC1=CC(=C(C=C1)C=C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -64.0°C
Density 0.9060g/mL
Boiling Point 73.0°C to 74.0°C (10.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C6H3CH=CH2
Refractive Index 1.5410 to 1.5440
Quantity 25 g
Specific Gravity 0.906
Stabilizer 0.05 % (ca.) t-butylcatechol
Formula Weight 132.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylstyrene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethylstyrene, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'