missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylanisole, 99%, ACROS Organics™

Brand:  Acros Organics 160001000

Code : 29

Additional Details : CAS Number : 6738-23-4 Weight : 0.15000kg

Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 10428160

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2, 4-Dimethylanisole
6738-23-4
100.0
99%
(CH3)2C6H3OCH3
MFCD00008377
99%
98.5
≥98.5%
C9H12O
136.19
1, 3-Dimethyl-4-methoxybenzene

Specifications

2, 4-Dimethylanisole
Clear colorless
99%
98.5
Authentic
1.5130 to 1.5150 (20°C, 589nm)
≥98.5%
C9H12O
136.19
06, 486
0.9700g/mL
100g
6738-23-4
100.0
Glass bottle
Liquid
99%
(CH3)2C6H3OCH3
MFCD00008377
1, 3-Dimethyl-4-methoxybenzene
Videos
SDS