missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide, ≥99%, Thermo Scientific™

Catalog Number p-7052650
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
3785-01-1
C17H21IN2
380.27
MFCD00011994
AMAXNNVXIBDEMV-UHFFFAOYSA-M
pinaflavol, daspei, 2-p-dimethylaminostyryl-1-ethylpyridinium iodide, 2-p-dimethylamino styryl ethyl pyridinium iodide, 2-4-dimethylamino styryl-1-ethylpyridinium iodide, 2-4-dimethylaminostyryl-1-ethylpyridinium iodide, pyridinium, 2-p-dimethylamino styryl-1-ethyl-, iodide, 2-2-4-dimethylamino phenyl ethenyl-1-ethylpyridinium iodide, 2-2-4-dimethylamino phenyl vinyl-1-ethylpyridinium iodide, pyridinium, 2-2-4-dimethylamino phenyl ethenyl-1-ethyl-, iodide
5713123
CHEBI:38007
4-[(E)-2-(1-ethylpyridin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide
This item is not returnable. View return policy
3785-01-1
C17H21IN2
380.27
MFCD00011994
AMAXNNVXIBDEMV-UHFFFAOYSA-M
pinaflavol, daspei, 2-p-dimethylaminostyryl-1-ethylpyridinium iodide, 2-p-dimethylamino styryl ethyl pyridinium iodide, 2-4-dimethylamino styryl-1-ethylpyridinium iodide, 2-4-dimethylaminostyryl-1-ethylpyridinium iodide, pyridinium, 2-p-dimethylamino styryl-1-ethyl-, iodide, 2-2-4-dimethylamino phenyl ethenyl-1-ethylpyridinium iodide, 2-2-4-dimethylamino phenyl vinyl-1-ethylpyridinium iodide, pyridinium, 2-2-4-dimethylamino phenyl ethenyl-1-ethyl-, iodide
5713123
CHEBI:38007
4-[(E)-2-(1-ethylpyridin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 267°C (decomposition)
Color Red
Quantity 250 mg
Beilstein 3783243
Sensitivity Light sensitive
SMILES CC[N+]1=CC=CC=C1C=CC2=CC=C(C=C2)N(C)C.[I-]
Formula Weight 380.27
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'