missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-[4-(Dimethylamino)phenyl]-5-nitrobenzimidazole, 95%

Brand:  Alfa Aesar™ H64145.03

Code : NEW

Additional Details : CAS Number : 111114-79-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468726

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

2-[4-(Dimethylamino)phenyl]-5-nitrobenzimidazole
282.3
N,N-Dimethyl-4-(5-nitro-2-benzimidazolyl)aniline
111114-79-5
MFCD00412131

Specifications

2-[4-(Dimethylamino)phenyl]-5-nitrobenzimidazole
111114-79-5
95%
MFCD00412131
1g
Solid
282.3
N,N-Dimethyl-4-(5-nitro-2-benzimidazolyl)aniline
Videos
SDS