missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-[4-(Dimethylamino)phenyl]-5-nitrobenzimidazole, 95%

Brand:  Alfa Aesar™ H64145.06

Code : NEW

Additional Details : CAS Number : 111114-79-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478726

  • £597.84 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-[4-(Dimethylamino)phenyl]-5-nitrobenzimidazole
282.3
N,N-Dimethyl-4-(5-nitro-2-benzimidazolyl)aniline
111114-79-5
MFCD00412131

Specifications

2-[4-(Dimethylamino)phenyl]-5-nitrobenzimidazole
111114-79-5
95%
MFCD00412131
5g
Solid
282.3
N,N-Dimethyl-4-(5-nitro-2-benzimidazolyl)aniline
Videos
SDS