missing translation for 'onlineSavingsMsg'
Learn More

(2,4-dimethyl-3-furyl)methylamine, Maybridge

Brand:  Maybridge CC65813DA

Code : 31

Additional Details : CAS Number : 1023814-75-6 Weight : 0.00100kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10118224

  • £411.00 / 1g


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Chemical Identifiers

(2, 4-dimethyl-3-furyl)methylamine
95%
C7H11NO
125.17

Specifications

(2, 4-dimethyl-3-furyl)methylamine
1023814-75-6
Amber Glass Bottle
125.17
1g
95%
C7H11NO
Videos
SDS