missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-ethylpyrrole, 96%, Thermo Scientific Chemicals

Catalog Number p-117232
Click to view available options
Quantity:
5 g
25 g
517-22-6
MFCD00005222
ZEBBLOXDLGIMEG-UHFFFAOYSA-N
2,4-dimethyl-3-ethylpyrrole, kryptopyrrole, kryptopyrrol, 3-ethyl-2,4-dimethylpyrrole, cryptopyrrole, 2,4-dimethyl-3-ethyl-1h-pyrrole, 1h-pyrrole, 3-ethyl-2,4-dimethyl, pyrrole, 3-ethyl-2,4-dimethyl, acmc-209kvh, 5-20-05-00099 beilstein handbook reference
10600
3-ethyl-2,4-dimethyl-1H-pyrrole
CCC1=C(NC=C1C)C
517-22-6
MFCD00005222
ZEBBLOXDLGIMEG-UHFFFAOYSA-N
2,4-dimethyl-3-ethylpyrrole, kryptopyrrole, kryptopyrrol, 3-ethyl-2,4-dimethylpyrrole, cryptopyrrole, 2,4-dimethyl-3-ethyl-1h-pyrrole, 1h-pyrrole, 3-ethyl-2,4-dimethyl, pyrrole, 3-ethyl-2,4-dimethyl, acmc-209kvh, 5-20-05-00099 beilstein handbook reference
10600
3-ethyl-2,4-dimethyl-1H-pyrrole
CCC1=C(NC=C1C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Density 0.9130g/mL
Boiling Point 197°C (710.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C8H13N
Refractive Index 1.4960 to 1.498
Quantity 5 g
Beilstein 20, I, 47
Specific Gravity 0.913
Molecular Weight (g/mol) 123.2
Formula Weight 123.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethyl-3-ethylpyrrole, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dimethyl-3-ethylpyrrole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'