missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1,3-thiazole-5-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number 10364802
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
80466-80-4
C5H6ClNO2S2
211.678
MFCD00052746
GFFJSTHQILQFNQ-UHFFFAOYSA-N
2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, dimethyl-1,3-thiazole-5-sulfonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 5-thiazolesulfonylchloride, 2,4-dimethyl, 2,4-dimethylthiazol-5-ylsulphonyl chloride
2776248
2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride
CC1=C(SC(=N1)C)S(=O)(=O)Cl
This item is not returnable. View return policy
80466-80-4
C5H6ClNO2S2
211.678
MFCD00052746
GFFJSTHQILQFNQ-UHFFFAOYSA-N
2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, dimethyl-1,3-thiazole-5-sulfonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 5-thiazolesulfonylchloride, 2,4-dimethyl, 2,4-dimethylthiazol-5-ylsulphonyl chloride
2776248
2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride
CC1=C(SC(=N1)C)S(=O)(=O)Cl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 211.69
Percent Purity 97%
Chemical Name or Material 2, 4-Dimethyl-1, 3-thiazole-5-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'