missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzophenone, 99%, Thermo Scientific Chemicals

Catalog Number p-7021425
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
131-56-6
C13H10O3
214.22
MFCD00002277
ZXDDPOHVAMWLBH-UHFFFAOYSA-N
2,4-dihydroxybenzophenone, 2,4-dihydroxyphenyl phenyl methanone, benzophenone-1, benzoresorcinol, resbenzophenone, inhibitor dhbp, uvinul 400, advastab 48, uvistat 12, methanone, 2,4-dihydroxyphenyl phenyl
8572
CHEBI:34240
(2,4-dihydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O
This item is not returnable. View return policy
131-56-6
C13H10O3
214.22
MFCD00002277
ZXDDPOHVAMWLBH-UHFFFAOYSA-N
2,4-dihydroxybenzophenone, 2,4-dihydroxyphenyl phenyl methanone, benzophenone-1, benzoresorcinol, resbenzophenone, inhibitor dhbp, uvinul 400, advastab 48, uvistat 12, methanone, 2,4-dihydroxyphenyl phenyl
8572
CHEBI:34240
(2,4-dihydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O

Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 144°C to 147°C
Density 1.32
Boiling Point 194°C (1 mmHg)
Flash Point 125°C (257°F)
Quantity 100 g
Beilstein 1311566
Merck Index 14,1106
Solubility Information Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material 2,4-Dihydroxybenzophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'